Merck
  • Muscarinic receptors promote castration-resistant growth of prostate cancer through a FAK-YAP signaling axis.
Muscarinic receptors promote castration-resistant growth of prostate cancer through a FAK-YAP signaling axis.
Oncogene (2020-03-25)
Yusuke Goto, Toshinori Ando, Hiroki Izumi, Xiaodong Feng, Nadia Arang, Mara Gilardi, Zhiyong Wang, Kazuyo Ando, J Silvio Gutkind
ABSTRACT
Prostate cancer (PCa) innervation contributes to the progression of PCa. However, the precise impact of innervation on PCa cells is still poorly understood. By focusing on muscarinic receptors, which are activated by the nerve-derived neurotransmitter acetylcholine, we show that muscarinic receptors 1 and 3 (m1 and m3) are highly expressed in PCa clinical specimens compared with all other cancer types, and that amplification or gain of their corresponding encoding genes (CHRM1 and CHRM3, respectively) represent a worse prognostic factor for PCa progression free survival. Moreover, m1 and m3 gene gain or amplification is frequent in castration-resistant PCa (CRPC) compared with hormone-sensitive PCa (HSPC) specimens. This was reflected in HSPC-derived cells, which show aberrantly high expression of m1 and m3 under androgen deprivation mimicking castration and androgen receptor inhibition. We also show that pharmacological activation of m1 and m3 signaling is sufficient to induce the castration-resistant growth of PCa cells. Mechanistically, we found that m1 and m3 stimulation induces YAP activation through FAK, whose encoding gene, PTK2 is frequently amplified in CRPC cases. Pharmacological inhibition of FAK and knockdown of YAP abolished m1 and m3-induced castration-resistant growth of PCa cells. Our findings provide novel therapeutic opportunities for muscarinic-signal-driven CRPC progression by targeting the FAK-YAP signaling axis.
Sigma-Aldrich
DL-Homocystine-1,1′-13C2, 99 atom % 13C
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Thrombin, Human Plasma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Clozapine N-oxide
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleoyl-L-α-lysophosphatidic acid sodium salt, ≥98%, solid
Sigma-Aldrich
Thrombin, Bovine, High Activity