Merck
  • Blockade of Oncogenic NOTCH1 with the SERCA Inhibitor CAD204520 in T Cell Acute Lymphoblastic Leukemia.
Blockade of Oncogenic NOTCH1 with the SERCA Inhibitor CAD204520 in T Cell Acute Lymphoblastic Leukemia.
Cell chemical biology (2020-05-11)
Matteo Marchesini, Andrea Gherli, Anna Montanaro, Laura Patrizi, Claudia Sorrentino, Luca Pagliaro, Chiara Rompietti, Samuel Kitara, Sabine Heit, Claus E Olesen, Jesper V Møller, Monia Savi, Leonardo Bocchi, Rocchina Vilella, Federica Rizzi, Marilena Baglione, Giorgia Rastelli, Caterina Loiacono, Roberta La Starza, Cristina Mecucci, Kimberly Stegmaier, Franco Aversa, Donatella Stilli, Anne-Marie Lund Winther, Paolo Sportoletti, Maike Bublitz, William Dalby-Brown, Giovanni Roti
ABSTRACT
The identification of SERCA (sarco/endoplasmic reticulum calcium ATPase) as a target for modulating gain-of-function NOTCH1 mutations in Notch-dependent cancers has spurred the development of this compound class for cancer therapeutics. Despite the innate toxicity challenge associated with SERCA inhibition, we identified CAD204520, a small molecule with better drug-like properties and reduced off-target Ca2+ toxicity compared with the SERCA inhibitor thapsigargin. In this work, we describe the properties and complex structure of CAD204520 and show that CAD204520 preferentially targets mutated over wild-type NOTCH1 proteins in T cell acute lymphoblastic leukemia (T-ALL) and mantle cell lymphoma (MCL). Uniquely among SERCA inhibitors, CAD204520 suppresses NOTCH1-mutated leukemic cells in a T-ALL xenografted model without causing cardiac toxicity. This study supports the development of SERCA inhibitors for Notch-dependent cancers and extends their application to cases with isolated mutations in the PEST degradation domain of NOTCH1, such as MCL or chronic lymphocytic leukemia (CLL).
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
L-(−)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Claycomb Medium, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%