Merck
  • T-2 toxin-induced apoptosis involving Fas, p53, Bcl-xL, Bcl-2, Bax and caspase-3 signaling pathways in human chondrocytes.
T-2 toxin-induced apoptosis involving Fas, p53, Bcl-xL, Bcl-2, Bax and caspase-3 signaling pathways in human chondrocytes.
Journal of Zhejiang University. Science. B (2008-06-11)
Jing-hong Chen, Jun-ling Cao, Yong-lie Chu, Zhi-lun Wang, Zhan-tian Yang, Hong-lin Wang
ABSTRACT
To investigate the effects of T-2 toxin on expressions of Fas, p53, Bcl-xL, Bcl-2, Bax and caspase-3 on human chondrocytes. Human chondrocytes were treated with T-2 toxin (1-20 ng/ml) for 5 d. Fas, p53 and other apoptosis-related proteins such as Bax, Bcl-2, Bcl-xL, caspase-3 were determined by Western blot analysis and their mRNA expressions were determined by reverse transcriptase-polymerase chain reaction (RT-PCR). Increases in Fas, p53 and the pro-apoptotic factor Bax protein and mRNA expressions and a decrease of the anti-apoptotic factor Bcl-xL were observed in a dose-dependent manner after exposures to 1-20 ng/ml T-2 toxin, while the expression of the anti-apoptotic factor Bcl-2 was unchanged. Meanwhile, T-2 toxin could also up-regulate the expressions of both pro-caspase-3 and caspase-3 in a dose-dependent manner. These data suggest a possible underlying molecular mechanism for T-2 toxin to induce the apoptosis signaling pathway in human chondrocytes by regulation of apoptosis-related proteins.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid