Merck
  • Induction of a Timed Metabolic Collapse to Overcome Cancer Chemoresistance.
Induction of a Timed Metabolic Collapse to Overcome Cancer Chemoresistance.
Cell metabolism (2020-08-09)
Nick van Gastel, Jessica B Spinelli, Azeem Sharda, Amir Schajnovitz, Ninib Baryawno, Catherine Rhee, Toshihiko Oki, Eliane Grace, Heather J Soled, Jelena Milosevic, David B Sykes, Peggy P Hsu, Matthew G Vander Heiden, Charles Vidoudez, Sunia A Trauger, Marcia C Haigis, David T Scadden
ABSTRACT
Cancer relapse begins when malignant cells pass through the extreme metabolic bottleneck of stress from chemotherapy and the byproducts of the massive cell death in the surrounding region. In acute myeloid leukemia, complete remissions are common, but few are cured. We tracked leukemia cells in vivo, defined the moment of maximal response following chemotherapy, captured persisting cells, and conducted unbiased metabolomics, revealing a metabolite profile distinct from the pre-chemo growth or post-chemo relapse phase. Persisting cells used glutamine in a distinctive manner, preferentially fueling pyrimidine and glutathione generation, but not the mitochondrial tricarboxylic acid cycle. Notably, malignant cell pyrimidine synthesis also required aspartate provided by specific bone marrow stromal cells. Blunting glutamine metabolism or pyrimidine synthesis selected against residual leukemia-initiating cells and improved survival in leukemia mouse models and patient-derived xenografts. We propose that timed cell-intrinsic or niche-focused metabolic disruption can exploit a transient vulnerability and induce metabolic collapse in cancer cells to overcome chemoresistance.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human GOT2
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, powder
Sigma-Aldrich
Poly(ethylene glycol), average Mn 400
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, powder
Sigma-Aldrich
BPTES, ≥95% (HPLC)
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution