Merck
  • p53 positively regulates osteoprotegerin-mediated inhibition of osteoclastogenesis by downregulating TSC2-induced autophagy in vitro.
p53 positively regulates osteoprotegerin-mediated inhibition of osteoclastogenesis by downregulating TSC2-induced autophagy in vitro.
Differentiation; research in biological diversity (2020-08-11)
Xishuai Tong, Jianhong Gu, Miaomiao Chen, Tao Wang, Hui Zou, Ruilong Song, Hongyan Zhao, Jianchun Bian, Zongping Liu
ABSTRACT
Osteoclasts are terminally multinucleated cells that are regulated by nuclear factor-activated T cells c1 (NFATc1), and are responsible for bone resorption while the tartrate resistant acid phosphatase (TRAP) enzymes releases into bone resorption lacunae. Furthermore, tumor suppressor p53 is a negative regulator during osteoclastogenesis. Osteoprotegerin (OPG) inhibits osteoclastogenesis and bone resorption by activating autophagy, however, whether p53 is involved in OPG-mediated inhibition of osteoclastogenesis remains unclear. In the current study, OPG could enhance the expression of p53 and tuberin sclerosis complex 2 (TSC2). Moreover, the expression of p53 is regulated by autophagy during OPG-mediated inhibition of osteoclastogenesis. Inhibition of p53 by treated with pifithrin-α (PFTα) causing augments of osteoclastogenesis and bone resorption, also reversed OPG-mediated inhibition of osteoclastogenesis by reducing the expression of TSC2. In addition, knockdown of TSC2 using siRNA could rescue OPG-mediated inhibition of osteoclastogenesis by reducing autophagy, which is manifested by the decrease of the expression of Beclin1 and the phosphorylation of mammalian target of rapamycin (mTOR) and ribosomal protein S6 kinase beta 1 (S6K1, also known as p70S6K). Collectively, p53 plays a critical role during OPG-mediated inhibition of osteoclastogenesis via regulating the TSC2-induced autophagy in vitro.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus