Merck
  • Defective mitochondrial ISCs biogenesis switches on IRP1 to fine tune selective mitophagy.
Defective mitochondrial ISCs biogenesis switches on IRP1 to fine tune selective mitophagy.
Redox biology (2020-08-17)
Hao Wu, Huifang Wei, Di Zhang, Sheikh Arslan Sehgal, Dejiu Zhang, Xiaohui Wang, Yan Qin, Lei Liu, Quan Chen
ABSTRACT
Both iron metabolism and mitophagy, a selective mitochondrial degradation process via autolysosomal pathway, are fundamental for the cellular well-being. Mitochondria are the major site for iron metabolism, especially the biogenesis of iron-sulfur clusters (ISCs) via the mitochondria-localized ISCs assembly machinery. Here we report that mitochondrial ISCs biogenesis is coupled with receptor-mediated mitophagy in mammalian cells. Perturbation of mitochondrial ISCs biogenesis, either by depleting iron with the iron chelator or by knocking down the core components of the mitochondrial ISCs assembly machinery, triggers FUNDC1-dependent mitophagy. IRP1, one of the cellular iron sensors to maintain iron homeostasis, is crucial for iron stresses induced mitophagy. Knockdown of IRP1 disturbed iron stresses induced mitophagy. Furthermore, IRP1 could bind to a newly characterized IRE in the 5' untranslated region of the Bcl-xL mRNA and suppress its translation. Bcl-xL is an intrinsic inhibitory protein of the mitochondrial phosphatase PGAM5, which catalyzes the dephosphorylation of FUNDC1 for mitophagy activation. Alterations of the IRP1/Bcl-xL axis navigate iron stresses induced mitophagy. We conclude that ISCs serve as physiological signals for mitophagy activation, thus coupling mitophagy with iron metabolism.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Anti-LYRM4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Anti-γ-Actin antibody, Mouse monoclonal, clone 2-2.1.14.17, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert