Merck
  • Isolation and culture of endothelial cells, pericytes and perivascular resident macrophage-like melanocytes from the young mouse ear.
Isolation and culture of endothelial cells, pericytes and perivascular resident macrophage-like melanocytes from the young mouse ear.
Nature protocols (2013-03-16)
Lingling Neng, Wenjing Zhang, Ahmed Hassan, Marcin Zemla, Allan Kachelmeier, Anders Fridberger, Manfred Auer, Xiaorui Shi
ABSTRACT
This protocol describes a growth medium-based approach for obtaining cochlear endothelial cells (ECs), pericytes (PCs) and perivascular resident macrophage-like melanocytes (PVM/Ms) from the stria vascularis of mice aged between P10 and P15 (P, postnatal day). The procedure does not involve mechanical or enzymatic digestion of the sample tissue. Explants of stria vascularis, 'mini-chips', are selectively cultured in growth medium, and primary cell lines are obtained in 7-10 d. The method is simple and reliable, and it provides high-quality ECs, PVM/Ms and PCs with a purity >90% after two passages. This protocol is suitable for producing primary culture cells from organs and tissues of small volume and high anatomical complexity, such as the inner ear capillaries. The highly purified primary cell lines enable cell culture-based in vitro modeling of cell-cell interactions, barrier control function and drug action.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Goat serum
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)