Merck
  • Clinical and functional characterization of recurrent missense variants implicated in THOC6-related intellectual disability.
Clinical and functional characterization of recurrent missense variants implicated in THOC6-related intellectual disability.
Human molecular genetics (2018-11-27)
Francesca Mattioli, Bertrand Isidor, Omar Abdul-Rahman, Andrew Gunter, Lijia Huang, Raman Kumar, Chandree Beaulieu, Jozef Gecz, Micheil Innes, Jean-Louis Mandel, Amélie Piton
ABSTRACT
THOC6 encodes a subunit of the THO complex that is part of a highly conserved transcription and export complex known to have roles in mRNA processing and export. Few homozygous or compound heterozygous variants have been identified in the THOC6 gene in patients with a syndromic form of intellectual disability [Beaulieu-Boycott-Innes syndrome (BBIS); MIM: 613680]. Here we report two additional individuals affected with BBIS originating from the north of Europe and sharing a haplotype composed of three very rare missense changes in the THOC6 gene-Trp100Arg, Val234Leu, Gly275Asp. The first individual is a boy who is homozygous for the three-variant haplotype due to a maternal uniparental disomy event. The second is a girl who is compound heterozygous for this haplotype and a previously reported Gly190Glu missense variant. We analyzed the impact of these different amino acid changes on THOC6 protein expression, cellular localization and interaction with the other THO complex subunits. We show that the different THOC6 variants alter the physiological nuclear localizationof the protein and its interaction with at least two THO subunits, THOC1 and THOC5. Two amino acid changes from the three-variant haplotype alone have specific effects and might contribute to the pathogenicity of the haplotype. Overall, we expanded the cohort of currently known individuals with BBIS by reporting two individuals carrying the same recurrent European haplotype composed of three amino acid changes, affecting THOC6 localization and interaction with THO protein partners.
Sigma-Aldrich
Anti-THOC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)