Merck
  • WNT11-FZD7-DAAM1 signalling supports tumour initiating abilities and melanoma amoeboid invasion.
WNT11-FZD7-DAAM1 signalling supports tumour initiating abilities and melanoma amoeboid invasion.
Nature communications (2020-10-22)
Irene Rodriguez-Hernandez, Oscar Maiques, Leonie Kohlhammer, Gaia Cantelli, Anna Perdrix-Rosell, Joanne Monger, Bruce Fanshawe, Victoria L Bridgeman, Sophia N Karagiannis, Rosa M Penin, Joaquim Marcolval, Rosa M Marti, Xavier Matias-Guiu, Gilbert O Fruhwirth, Jose L Orgaz, Ilaria Malanchi, Victoria Sanz-Moreno
ABSTRACT
Melanoma is a highly aggressive tumour that can metastasize very early in disease progression. Notably, melanoma can disseminate using amoeboid invasive strategies. We show here that high Myosin II activity, high levels of ki-67 and high tumour-initiating abilities are characteristic of invasive amoeboid melanoma cells. Mechanistically, we find that WNT11-FZD7-DAAM1 activates Rho-ROCK1/2-Myosin II and plays a crucial role in regulating tumour-initiating potential, local invasion and distant metastasis formation. Importantly, amoeboid melanoma cells express both proliferative and invasive gene signatures. As such, invasive fronts of human and mouse melanomas are enriched in amoeboid cells that are also ki-67 positive. This pattern is further enhanced in metastatic lesions. We propose eradication of amoeboid melanoma cells after surgical removal as a therapeutic strategy.
Sigma-Aldrich
(±)-Blebbistatin, (±)-Blebbistatin, CAS 674289-55-5, is a cell-permeable, selective, and reversible inhibitor of nonmuscle myosin II. Blocks cell blebbing and disrupts cell migration & cytoKinesis in vertebrate cells.
Sigma-Aldrich
Rho Kinase Inhibitor, The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse