Merck
  • CHD7 and 53BP1 regulate distinct pathways for the re-ligation of DNA double-strand breaks.
CHD7 and 53BP1 regulate distinct pathways for the re-ligation of DNA double-strand breaks.
Nature communications (2020-11-15)
Magdalena B Rother, Stefania Pellegrino, Rebecca Smith, Marco Gatti, Cornelia Meisenberg, Wouter W Wiegant, Martijn S Luijsterburg, Ralph Imhof, Jessica A Downs, Alfred C O Vertegaal, Sébastien Huet, Matthias Altmeyer, Haico van Attikum
ABSTRACT
Chromatin structure is dynamically reorganized at multiple levels in response to DNA double-strand breaks (DSBs). Yet, how the different steps of chromatin reorganization are coordinated in space and time to differentially regulate DNA repair pathways is insufficiently understood. Here, we identify the Chromodomain Helicase DNA Binding Protein 7 (CHD7), which is frequently mutated in CHARGE syndrome, as an integral component of the non-homologous end-joining (NHEJ) DSB repair pathway. Upon recruitment via PARP1-triggered chromatin remodeling, CHD7 stimulates further chromatin relaxation around DNA break sites and brings in HDAC1/2 for localized chromatin de-acetylation. This counteracts the CHD7-induced chromatin expansion, thereby ensuring temporally and spatially controlled 'chromatin breathing' upon DNA damage, which we demonstrate fosters efficient and accurate DSB repair by controlling Ku and LIG4/XRCC4 activities. Loss of CHD7-HDAC1/2-dependent cNHEJ reinforces 53BP1 assembly at the damaged chromatin and shifts DSB repair to mutagenic NHEJ, revealing a backup function of 53BP1 when cNHEJ fails.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Romidepsin, ≥98% (HPLC)
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography