Merck
  • Small-Molecule MYC Inhibitors Suppress Tumor Growth and Enhance Immunotherapy.
Small-Molecule MYC Inhibitors Suppress Tumor Growth and Enhance Immunotherapy.
Cancer cell (2019-11-05)
Huiying Han, Atul D Jain, Mihai I Truica, Javier Izquierdo-Ferrer, Jonathan F Anker, Barbara Lysy, Vinay Sagar, Yi Luan, Zachary R Chalmers, Kenji Unno, Hanlin Mok, Rajita Vatapalli, Young A Yoo, Yara Rodriguez, Irawati Kandela, J Brandon Parker, Debabrata Chakravarti, Rama K Mishra, Gary E Schiltz, Sarki A Abdulkadir
ABSTRACT
Small molecules that directly target MYC and are also well tolerated in vivo will provide invaluable chemical probes and potential anti-cancer therapeutic agents. We developed a series of small-molecule MYC inhibitors that engage MYC inside cells, disrupt MYC/MAX dimers, and impair MYC-driven gene expression. The compounds enhance MYC phosphorylation on threonine-58, consequently increasing proteasome-mediated MYC degradation. The initial lead, MYC inhibitor 361 (MYCi361), suppressed in vivo tumor growth in mice, increased tumor immune cell infiltration, upregulated PD-L1 on tumors, and sensitized tumors to anti-PD1 immunotherapy. However, 361 demonstrated a narrow therapeutic index. An improved analog, MYCi975 showed better tolerability. These findings suggest the potential of small-molecule MYC inhibitors as chemical probes and possible anti-cancer therapeutic agents.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
10074-G5, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Pricing and availability is not currently available.
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Pricing and availability is not currently available.