Merck
  • Nox4 regulates InsP3 receptor-dependent Ca2+ release into mitochondria to promote cell survival.
Nox4 regulates InsP3 receptor-dependent Ca2+ release into mitochondria to promote cell survival.
The EMBO journal (2020-10-02)
Matteo Beretta, Celio Xc Santos, Chris Molenaar, Anne D Hafstad, Chris Cj Miller, Aram Revazian, Kai Betteridge, Katrin Schröder, Katrin Streckfuß-Bömeke, James H Doroshow, Roland A Fleck, Tsung-Ping Su, Vsevolod V Belousov, Maddy Parsons, Ajay M Shah
ABSTRACT
Cells subjected to environmental stresses undergo regulated cell death (RCD) when homeostatic programs fail to maintain viability. A major mechanism of RCD is the excessive calcium loading of mitochondria and consequent triggering of the mitochondrial permeability transition (mPT), which is especially important in post-mitotic cells such as cardiomyocytes and neurons. Here, we show that stress-induced upregulation of the ROS-generating protein Nox4 at the ER-mitochondria contact sites (MAMs) is a pro-survival mechanism that inhibits calcium transfer through InsP3 receptors (InsP3 R). Nox4 mediates redox signaling at the MAM of stressed cells to augment Akt-dependent phosphorylation of InsP3 R, thereby inhibiting calcium flux and mPT-dependent necrosis. In hearts subjected to ischemia-reperfusion, Nox4 limits infarct size through this mechanism. These results uncover a hitherto unrecognized stress pathway, whereby a ROS-generating protein mediates pro-survival effects through spatially confined signaling at the MAM to regulate ER to mitochondria calcium flux and triggering of the mPT.
Sigma-Aldrich
Human TNNI3 / Troponin I, Cardiac Muscle ELISA Kit
Sigma-Aldrich
MISSION® esiRNA, targeting human UCK2
Sigma-Aldrich
Xestospongin C, film
Supelco
Malachite Green chloride, analytical standard