Merck
UDP-glucuronosyltransferases and clinical drug-drug interactions.
Pharmacology & therapeutics (2005-03-23)
Tony K L Kiang, Mary H H Ensom, Thomas K H Chang
ABSTRACT
UDP-glucuronosyltransferase (UGT) enzymes catalyze the conjugation of various endogenous substances (e.g., bilirubin) and exogenous compounds (e.g., drugs). The human UGT superfamily is comprised of 2 families (UGT1 and UGT2) and 3 subfamilies (UGT1A, UGT2A, and UGT2B). Many of the individual UGT enzymes are expressed not only in liver but also in extrahepatic tissues, where the extent of glucuronidation can be substantial. Several others (e.g., UGT1A7, UGT1A8, and UGT1A10) are expressed only in extrahepatic tissues. The molecular regulation of UGT enzyme is still not fully understood, but various transcription factors appear to play a regulatory role. The expression of individual UGT enzymes is subject to genetic polymorphism and these enzymes can be inhibited or induced by xenobiotics. Experimental evidence in humans indicates that the glucuronidation of acetaminophen, codeine, zidovudine, carbamazepine, lorazepam, and propafenone can influenced by specific interacting drugs. In contrast, the glucuronidation of diflunisal, morphine, naproxen, and temazepam is not affected appreciably by the drugs investigated to date. In general, UGT-mediated human drug interaction studies are difficult to interpret. The factors that complicate the interpretation of this type of drug interaction data are discussed.
Product Description
Supelco
Ibuprofen
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Phenylbutazone
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%