Merck
  • Docking and three-dimensional quantitative structure-activity relationship (3D QSAR) analyses of nonsteroidal progesterone receptor ligands.
Docking and three-dimensional quantitative structure-activity relationship (3D QSAR) analyses of nonsteroidal progesterone receptor ligands.
Journal of medicinal chemistry (2006-07-11)
Annu A Söderholm, Pekka T Lehtovuori, Tommi H Nyrönen
ABSTRACT
We report a docking and comparative molecular similarity indices analysis (CoMSIA) study of progesterone receptor (PR) ligands with an emphasis on nonsteroids including tanaproget. The ligand alignment generation, a critical part of model building, comprised two stages. First, thorough conformational sampling of docking poses within the PR binding pocket was made with the program GOLD. Second, a strategy to select representative poses for CoMSIA was developed utilizing the FlexX scoring function. After manual replacement of five poses where this approach had problems, a significant correlation (r(2) = 0.878) between the experimental affinities and electrostatic, hydrophobic, and hydrogen bond donor properties of the aligned ligands was found. Extensive model validation was made using random-group cross-validations, external test set predictions (r(pred)(2) = 0.833), and consistency check between the CoMSIA model and the PR binding site structure. Robustness, predictive ability, and automated alignment generation make the model a potential tool for virtual screening.
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture