Merck
  • Carbonic anhydrase activators: the first X-ray crystallographic study of an adduct of isoform I.
Carbonic anhydrase activators: the first X-ray crystallographic study of an adduct of isoform I.
Bioorganic & medicinal chemistry letters (2006-07-28)
Claudia Temperini, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The X-ray crystallographic structure for the adduct of an activator with human carbonic anhydrase isozyme I (hCA I) is reported. L-Histidine binds deep within the enzyme active site, participating in a network of hydrogen bonds involving its carboxylate moiety and the zinc-bound water molecule, as well as the imidazole of His200, being in van der Waals contacts with Thr199, His200, His64, and His67. This binding is very different from that to the other major cytosolic isozyme hCA II.
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland