Merck
  • Analogues of acifran: agonists of the high and low affinity niacin receptors, GPR109a and GPR109b.
Analogues of acifran: agonists of the high and low affinity niacin receptors, GPR109a and GPR109b.
Journal of medicinal chemistry (2007-03-16)
Jae-Kyu Jung, Benjamin R Johnson, Tracy Duong, Marc Decaire, Jane Uy, Tawfik Gharbaoui, P Douglas Boatman, Carleton R Sage, Ruoping Chen, Jeremy G Richman, Daniel T Connolly, Graeme Semple
ABSTRACT
Recently identified GPCRs, GPR109a and GPR109b, the high and low affinity receptors for niacin, may represent good targets for the development of HDL elevating drugs for the treatment of atherosclerosis. Acifran, an agonist of both receptors, has been tested in human subjects, yet until recently very few analogs had been reported. We describe a series of acifran analogs prepared using newly developed synthetic pathways and evaluated as agonists for GPR109a and GPR109b, resulting in identification of compounds with improved activity at these receptors.
Product Description
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%