Merck
FMRP acts as a key messenger for visceral pain modulation.
Molecular pain (2020-11-28)
Liu-Kun Yang, Liang Lu, Ban Feng, Xin-Shang Wang, Jiao Yue, Xu-Bo Li, Min Zhuo, Shui-Bing Liu
ABSTRACT
Visceral pain is a common clinical symptom, which is caused by mechanical stretch, spasm, ischemia and inflammation. Fragile X syndrome (FXS) with lack of fragile X mental retardation protein (FMRP) protein is an inherited disorder that is characterized by moderate or severe intellectual and developmental disabilities. Previous studies reported that FXS patients have self-injurious behavior, which may be associated with deficits in nociceptive sensitization. However, the role of FMRP in visceral pain is still unclear. In this study, the FMR1 knock out (KO) mice and SH-SY5Y cell line were employed to demonstrate the role of FMRP in the regulation of visceral pain. The data showed that FMR1 KO mice were insensitive to zymosan treatment. Recording in the anterior cingulate cortex (ACC), a structure involved in pain process, showed less presynaptic glutamate release and postsynaptic responses in the FMR1 KO mice as compared to the wild type (WT) mice after zymosan injection. Zymosan treatment caused enhancements of adenylyl cyclase 1 (AC1), a pain-related enzyme, and NMDA GluN2B receptor in the ACC. However, these up-regulations were attenuated in the ACC of FMR1 KO mice. Last, we found that zymosan treatment led to increase of FMRP levels in the ACC. These results were further confirmed in SH-SY5Y cells in vitro. Our findings demonstrate that FMRP is required for NMDA GluN2B and AC1 upregulation, and GluN2B/AC1/FMRP forms a positive feedback loop to modulate visceral pain.
Sigma-Aldrich
Anti-phospho-NR2B (Ser1303) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-NR1 Antibody, CT, Upstate®, from mouse
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NMDAR2A Antibody, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-NMDAR2B Antibody, pain, serum, Chemicon®