Merck
  • Loss of TAX1BP1-Directed Autophagy Results in Protein Aggregate Accumulation in the Brain.
Loss of TAX1BP1-Directed Autophagy Results in Protein Aggregate Accumulation in the Brain.
Molecular cell (2020-11-19)
Shireen A Sarraf, Hetal V Shah, Gil Kanfer, Alicia M Pickrell, Lynne A Holtzclaw, Michael E Ward, Richard J Youle, Shireen A Sarraf, Hetal V Shah, Gil Kanfer, Alicia M Pickrell, Lynne A Holtzclaw, Michael E Ward, Richard J Youle
ABSTRACT
Protein aggregates disrupt cellular homeostasis, causing toxicity linked to neurodegeneration. Selective autophagic elimination of aggregates is critical to protein quality control, but how aggregates are selectively targeted for degradation is unclear. We compared the requirements for autophagy receptor proteins: OPTN, NBR1, p62, NDP52, and TAX1BP1 in clearance of proteotoxic aggregates. Endogenous TAX1BP1 is recruited to and required for the clearance of stress-induced aggregates, whereas ectopic expression of TAX1BP1 increases clearance through autophagy, promoting viability of human induced pluripotent stem cell-derived neurons. In contrast, TAX1BP1 depletion sensitizes cells to several forms of aggregate-induced proteotoxicity. Furthermore, TAX1BP1 is more specifically expressed in the brain compared to other autophagy receptor proteins. In vivo, loss of TAX1BP1 results in accumulation of high molecular weight ubiquitin conjugates and premature lipofuscin accumulation in brains of young TAX1BP1 knockout mice. TAX1BP1 mediates clearance of a broad range of cytotoxic proteins indicating therapeutic potential in neurodegenerative diseases.
Sigma-Aldrich
Anti-TAX1BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Poly-D-lysine hydrobromide, average mol wt 30,000-70,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Supelco
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid, pharmaceutical impurity standard
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)