Merck
  • Synthesis and antibacterial activities of new quinolone derivatives utilizing 1-azabicyclo[1.1.0]butane.
Synthesis and antibacterial activities of new quinolone derivatives utilizing 1-azabicyclo[1.1.0]butane.
Bioorganic & medicinal chemistry letters (2006-12-13)
Yoshifumi Ikee, Kana Hashimoto, Masaaki Nakashima, Kazuhiko Hayashi, Shigeki Sano, Motoo Shiro, Yoshimitsu Nagao
ABSTRACT
The ring-opening reactions of 1-azabicyclo[1.1.0]butane 3 with thiols 6a-f gave 3-sulfenylazetidine derivatives 7a-f in 50-92% yields. Treatment of 3 with aromatic amines 11a-e and dibenzylamine 11f in the presence of Mg(ClO(4))(2) afforded the corresponding 3-aminoazetidine derivatives 12a-f in 24-53% yields. These azetidine derivatives were introduced into the C7 position of a quinolone nucleus 8 to afford the corresponding fluoroquinolones 9a-f and 13a-f in 21-83% yields. Some of them exhibited superior antibacterial activity against quinolone-susceptible MRSA in comparison with clinically used fluoroquinolones, such as levofloxacin, ciprofloxacin, and gatifloxacin.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)