Merck
  • Caco-2 cell permeability and stability of two d-glucopyranuronamide conjugates of thyrotropin-releasing hormone.
Caco-2 cell permeability and stability of two d-glucopyranuronamide conjugates of thyrotropin-releasing hormone.
Bioorganic & medicinal chemistry (2007-05-15)
Susanne T Wessling, Benjamin P Ross, Yasuko Koda, Joanne T Blanchfield, Istvan Toth
ABSTRACT
Caco-2 cell permeability and stability assays were used as an in vitro model to study the intestinal epithelial transport and stability of two analogues of thyrotropin-releasing hormone (TRH; Pyr-His-Pro-NH2). Peptide 1 (Pyr-His-Pro-D-glucopyranuronamide) was more permeable across the Caco-2 cell monolayer compared with the permeability of the parent TRH peptide (Papp=5.10+/-1.89x10(-6) cm/s c.f. Papp=0.147+/-0.0474x10(-6) cm/s respectively). The permeability of peptide 1 was improved threefold by attaching a 2-aminooctanoic acid moiety to the N-terminus to form peptide 2 (2-aminooctanoic acid-Gln-His-Pro-D-glucopyranuronamide) (Papp=16.3+/-2.47x10(-6) cm/s). The half-life for both peptide 1 and peptide 2 was approximately 20 min in a homogenate of Caco-2 cells compared with the half-life of TRH which is approximately 3 min. It was concluded that the permeability of peptides 1 and 2 was enhanced because of their increased stability, while the higher permeability of peptide 2 compared with peptide 1 may be attributed to its increased lipophilicity which results in enhanced passive diffusion.
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Thyrotropin releasing hormone, ≥98% (HPLC), powder