Merck
  • CCL5-dependent mast cell infiltration into the tumor microenvironment in clear cell renal cell carcinoma patients.
CCL5-dependent mast cell infiltration into the tumor microenvironment in clear cell renal cell carcinoma patients.
Aging (2020-11-13)
Tianjie Liu, Qing Xia, Haibao Zhang, Zixi Wang, Wenjie Yang, Xiaoyun Gu, Tao Hou, Yule Chen, Xinqi Pei, Guodong Zhu, Dalin He, Lei Li, Shan Xu
ABSTRACT
We investigated the mechanisms affecting tumor progression and survival outcomes in Polybromo-1-mutated (PBRM1MUT) clear cell renal cell carcinoma (ccRCC) patients. PBRM1MUT ccRCC tissues contained higher numbers of mast cells and lower numbers of CD8+ and CD4+ T cells than tissues from PBRM1WT ccRCC patients. Hierarchical clustering, pathway enrichment and GSEA analyses demonstrated that PBRM1 mutations promote tumor progression by activating hypoxia inducible factor (HIF)-related signaling pathways and increasing expression of vascular endothelial growth factor family genes. PBRM1MUT ccRCC tissues also show increased expression of C-C motif chemokine ligand 5 (CCL5). PBRM1-silenced ccRCC cells exhibited greater Matrigel tube formation and cell proliferation than controls. In addition, HMC-1 human mast cells exhibited CCL5-dependent in vitro migration on Transwell plates. High CCL5 expression in PBRM1MUT ccRCC patients correlated with increased expression of genes encoding IFN-γ, IFN-α, IL-6, JAK-STAT3, TNF-α, and NF-ΚB. Moreover, high CCL5 expression was associated with poorer survival outcomes in ccRCC patients. These findings demonstrate that CCL5-dependent mast cell infiltration promotes immunosuppression within the tumor microenvironment, resulting in tumor progression and adverse survival outcomes in PBRM1MUT ccRCC patients.
Sigma-Aldrich
MISSION® esiRNA, targeting human PBRM1
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile