Merck
  • The Huntingtin-interacting protein SETD2/HYPB is an actin lysine methyltransferase.
The Huntingtin-interacting protein SETD2/HYPB is an actin lysine methyltransferase.
Science advances (2020-10-04)
Riyad N H Seervai, Rahul K Jangid, Menuka Karki, Durga Nand Tripathi, Sung Yun Jung, Sarah E Kearns, Kristen J Verhey, Michael A Cianfrocco, Bryan A Millis, Matthew J Tyska, Frank M Mason, W Kimryn Rathmell, In Young Park, Ruhee Dere, Cheryl Lyn Walker
ABSTRACT
The methyltransferase SET domain-containing 2 (SETD2) was originally identified as Huntingtin (HTT) yeast partner B. However, a SETD2 function associated with the HTT scaffolding protein has not been elucidated, and no linkage between HTT and methylation has yet been uncovered. Here, we show that SETD2 is an actin methyltransferase that trimethylates lysine-68 (ActK68me3) in cells via its interaction with HTT and the actin-binding adapter HIP1R. ActK68me3 localizes primarily to the insoluble F-actin cytoskeleton in cells and regulates actin polymerization/depolymerization dynamics. Disruption of the SETD2-HTT-HIP1R axis inhibits actin methylation, causes defects in actin polymerization, and impairs cell migration. Together, these data identify SETD2 as a previously unknown HTT effector regulating methylation and polymerization of actin filaments and provide new avenues for understanding how defects in SETD2 and HTT drive disease via aberrant cytoskeletal methylation.
Sigma-Aldrich
Jasplakinolide, ≥97% (HPLC)
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
SETD2 active human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)