Merck
  • Identification of new functional inhibitors of acid sphingomyelinase using a structure-property-activity relation model.
Identification of new functional inhibitors of acid sphingomyelinase using a structure-property-activity relation model.
Journal of medicinal chemistry (2007-11-22)
Johannes Kornhuber, Philipp Tripal, Martin Reichel, Lothar Terfloth, Stefan Bleich, Jens Wiltfang, Erich Gulbins
ABSTRACT
Some organic weak bases induce a detachment from inner lysosomal membranes and subsequent inactivation of acid sphingomyelinase (ASM) and thus work as functional ASM inhibitors. The aim of the present investigation was to develop a structure-property-activity relation (SPAR) model in order to specify the structural and physicochemical characteristics of probes capable of functionally inhibiting ASM. High p K a and high log P values are necessary but not sufficient preconditions for functional inhibition of ASM. The experimental data supported the requirement of an additional factor, which is necessary for functional inhibition of ASM. This factor k is related to the steric hindrance of the most basic nitrogen atom and presumably modulates the free presentation of a protonated nitrogen atom at the inner lysosomal surface. During the course of the study, we characterized 26 new functional ASM inhibitors, including doxepine 63, fluoxetine 104, maprotiline 109, nortriptyline 114, paroxetine 118, sertraline 124, suloctidil 125, and terfenadine 127.
Sigma-Aldrich
Harmine, 98%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
Demecolcine solution, 10 μg/mL in HBSS, ACF Qualified, BioXtra
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Ritanserin, powder
Sigma-Aldrich
Demecolcine, ≥98% (HPLC)
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
5-Azacytidine, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Terfenadine