Merck
  • Oncogenic herpesvirus KSHV triggers hallmarks of alternative lengthening of telomeres.
Oncogenic herpesvirus KSHV triggers hallmarks of alternative lengthening of telomeres.
Nature communications (2021-01-23)
Timothy P Lippert, Paulina Marzec, Aurora I Idilli, Grzegorz Sarek, Aleksandra Vancevska, Mark Bower, Paul J Farrell, Päivi M Ojala, Niklas Feldhahn, Simon J Boulton
ABSTRACT
To achieve replicative immortality, cancer cells must activate telomere maintenance mechanisms to prevent telomere shortening. ~85% of cancers circumvent telomeric attrition by re-expressing telomerase, while the remaining ~15% of cancers induce alternative lengthening of telomeres (ALT), which relies on break-induced replication (BIR) and telomere recombination. Although ALT tumours were first reported over 20 years ago, the mechanism of ALT induction remains unclear and no study to date has described a cell-based model that permits the induction of ALT. Here, we demonstrate that infection with Kaposi's sarcoma herpesvirus (KSHV) induces sustained acquisition of ALT-like features in previously non-ALT cell lines. KSHV-infected cells acquire hallmarks of ALT activity that are also observed in KSHV-associated tumour biopsies. Down-regulating BIR impairs KSHV latency, suggesting that KSHV co-opts ALT for viral functionality. This study uncovers KSHV infection as a means to study telomere maintenance by ALT and reveals features of ALT in KSHV-associated tumours.
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
DIG Oligonucleotide 3′-End Labeling Kit, 2nd generation, sufficient for 25 labeling reactions (100 pmol of oligonucleotides per assay; 1 ug of a 30-mer oligonucleotide), storage condition avoid repeated freeze/thaw cycles
Roche
Random Primed DNA Labeling Kit
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
SYBR® Green JumpStart Taq ReadyMix, for quantitative PCR, MgCI2 in buffer