Merck
  • Antimycobacterial and H1-antihistaminic activity of 2-substituted piperidine derivatives.
Antimycobacterial and H1-antihistaminic activity of 2-substituted piperidine derivatives.
Bioorganic & medicinal chemistry (2008-11-04)
Robert Weis, Klaus Schweiger, Johanna Faist, Erich Rajkovic, Andreas J Kungl, Walter M F Fabian, Walter Schunack, Werner Seebacher
ABSTRACT
2-Substituted derivatives of the antihistaminic agents Bamipine, Diphenylpyraline and of their 1-phenyl analogues were tested for their antimycobacterial and H(1)-antagonistic activities. They are strong H1-receptor antagonists and also inhibit the growth of mycobacterials with a maximum MIC of 6.25 microg/mL against Mycobacterium tuberculosis H(37)Rv. H1-receptor antagonistic potency was slightly decreased by substitution in ring position 2 and distinctly diminished by N-aryl substitution. The antimycobacterial potency of Diphenylpyraline was in general increased by substitution in ring position 2, whereas only a few Bamipine derivatives showed markedly improved activity. A correlation between the two activities was not detected for those compounds.
Product Description
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals