Merck
  • Multiple unbiased approaches identify oxidosqualene cyclase as the molecular target of a promising anti-leishmanial.
Multiple unbiased approaches identify oxidosqualene cyclase as the molecular target of a promising anti-leishmanial.
Cell chemical biology (2021-03-11)
Luciana S Paradela, Richard J Wall, Sandra Carvalho, Giulia Chemi, Victoriano Corpas-Lopez, Eoin Moynihan, Davide Bello, Stephen Patterson, Maria Lucia S Güther, Alan H Fairlamb, Michael A J Ferguson, Fabio Zuccotto, Julio Martin, Ian H Gilbert, Susan Wyllie
ABSTRACT
Phenotypic screening identified a benzothiophene compound with activity against Leishmania donovani, the causative agent of visceral leishmaniasis. Using multiple orthogonal approaches, oxidosqualene cyclase (OSC), a key enzyme of sterol biosynthesis, was identified as the target of this racemic compound and its enantiomers. Whole genome sequencing and screening of a genome-wide overexpression library confirmed that OSC gene amplification is associated with resistance to compound 1. Introduction of an ectopic copy of the OSC gene into wild-type cells reduced susceptibility to these compounds confirming the role of this enzyme in resistance. Biochemical analyses demonstrated the accumulation of the substrate of OSC and depletion of its product in compound (S)-1-treated-promastigotes and cell-free membrane preparations, respectively. Thermal proteome profiling confirmed that compound (S)-1 binds directly to OSC. Finally, modeling and docking studies identified key interactions between compound (S)-1 and the LdOSC active site. Strategies to improve the potency for this promising anti-leishmanial are proposed.
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(S)-GSK2920487A, ≥98% (HPLC)
Sigma-Aldrich
Posaconazole, ≥98% (HPLC)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
PCR DIG Probe Synthesis Kit, sufficient for 25 reaction (50 μL final reaction volume)