Merck
  • Inhibition of 15-lipoxygenase-catalysed oxygenation of arachidonic acid by substituted benzoic acids.
Inhibition of 15-lipoxygenase-catalysed oxygenation of arachidonic acid by substituted benzoic acids.
Bioorganic & medicinal chemistry (2008-03-04)
Wendy R Russell, Lorraine Scobbie, Garry G Duthie, Andrew Chesson
ABSTRACT
Elevated levels of phospholipases, prostaglandin synthases and lipoxygenases in colonic cells at various stages of malignancy indicate a strong link between dietary lipids and colon cancer. Lipoxygenase-catalysed arachidonic acid metabolism plays a key role in colorectal carcinogenesis and has the potential to be modulated by phenolic compounds. Plant-based foods are rich sources of phenolic compounds and in the human colon they are predominantly available as simple phenolics such as the benzoic acids. Benzoic acids were determined in faecal waters from four volunteers consuming a western-style diet. Structure-activity relationships were established for the lipoxygenase-catalysed oxygenation of arachidonic acid using an oxygen electrode. All compounds studied inhibited this reaction (21-73%; p<0.001) and many of the structural features could be rationalised by computational modelling. No correlation was observed with the ability to act as reductants, supporting the hypothesis that their mode of inhibition may not be by a direct redox effect on the non-haem iron.
Sigma-Aldrich
2,4-Dihydroxybenzoic acid, 97%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
3,5-Dihydroxybenzoic acid, 97%
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
2,6-Dihydroxybenzoic acid, 98%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%