Merck
  • Understanding existing and designing novel synthetic routes to Pd-PEPPSI-NHC and Pd-PEPPSI-PR3 pre-catalysts.
Understanding existing and designing novel synthetic routes to Pd-PEPPSI-NHC and Pd-PEPPSI-PR3 pre-catalysts.
Chemical communications (Cambridge, England) (2020-04-30)
Sébastien G Guillet, Vladislav A Voloshkin, Marina Saab, Marek Beliš, Kristof Van Hecke, Fady Nahra, Steven P Nolan
ABSTRACT
The reaction mechanism leading to the formation of cross-coupling palladium pre-catalysts of the PEPPSI family was investigated. Two intermediates were isolated and proved to be both suitable synthons to the pre-catalysts, with one permitting the design of a novel and greener user-friendly synthetic route. In light of this mechanistic understanding, the traditional one-pot method was shown to be possible using stoichiometric amounts of throw-away ligand, which represents a considerable synthetic improvement over the wasteful "in pyridine" approach.
Sigma-Aldrich
IPr*-PEPPSI, ≥95%
Sigma-Aldrich
[SIPr-H]2Pd2Cl6
Sigma-Aldrich
[IPr-H]2Pd2Cl6, ≥95%