Merck
  • Go protein subunit Goα and the secretory process of the natriuretic peptide hormones ANF and BNP.
Go protein subunit Goα and the secretory process of the natriuretic peptide hormones ANF and BNP.
Journal of molecular endocrinology (2015-04-29)
Cassandra Roeske, Amy Martinuk, Asna Choudhry, Geoffrey N Hendy, Michael Gollob, Qiuji Li, Tina Georgalis, Adolfo J de Bold
ABSTRACT
Expression of the G protein subunit Goα has been shown to be prominent in the atria of the rat heart and to be significantly associated with atrial natriuretic factor (ANF)-containing atrial-specific secretory granules by immunocytochemistry. In addition, differential expression profile analysis using oligonucleotide arrays has shown that the Goα isoform 1 (Goα1) is 2.3-fold more abundant in the atria than it is in the ventricles. In the present report, we show protein-protein interaction between Goα and ANF by yeast two-hybrid and by immunoprecipitation. A cardiac conditional Goα knockout model developed for the present study showed a 90% decrease in Goα expression and decreased atrial expression and ANF and brain natriuretic peptides (BNP) content. Expression of chromogranin A, a specific atrial granule core constituent, was not affected. Morphometric assessment of atrial tissue showed a very significant decrease in atrial-specific granule density as well as granule core electron density. Atrial electrical activity was not affected. The results obtained are compatible with the suggestion that Goα plays a role in ANF sorting during intracellular vectorial transport and with the presence of a mechanism that preserves the molar relationship between cellular ANF and BNP stores in the face of the decreased production of these hormones.
Sigma-Aldrich
REDExtract-N-Amp Tissue PCR Kit, sufficient for 10 reactions, sufficient for 100 reactions, sufficient for 1000 reactions, hotstart, dNTPs included
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution