Merck
  • αv-Class integrin binding to fibronectin is solely mediated by RGD and unaffected by an RGE mutation.
αv-Class integrin binding to fibronectin is solely mediated by RGD and unaffected by an RGE mutation.
The Journal of cell biology (2020-11-04)
María Benito-Jardón, Nico Strohmeyer, Sheila Ortega-Sanchís, Mitasha Bharadwaj, Markus Moser, Daniel J Müller, Reinhard Fässler, Mercedes Costell
ABSTRACT
Fibronectin (FN) is an essential glycoprotein of the extracellular matrix; binds integrins, syndecans, collagens, and growth factors; and is assembled by cells into complex fibrillar networks. The RGD motif in FN facilitates cell binding- and fibrillogenesis through binding to α5β1 and αv-class integrins. However, whether RGD is the sole binding site for αv-class integrins is unclear. Most notably, substituting aspartate with glutamate (RGE) was shown to eliminate integrin binding in vitro, while mouse genetics revealed that FNRGE preserves αv-class integrin binding and fibrillogenesis. To address this conflict, we employed single-cell force spectroscopy, engineered cells, and RGD motif-deficient mice (Fn1ΔRGD/ΔRGD) to search for additional αv-class integrin-binding sites. Our results demonstrate that α5β1 and αv-class integrins solely recognize the FN-RGD motif and that αv-class, but not α5β1, integrins retain FN-RGE binding. Furthermore, Fn1ΔRGD/ΔRGD tissues and cells assemble abnormal and dysfunctional FNΔRGD fibrils in a syndecan-dependent manner. Our data highlight the central role of FN-RGD and the functionality of FN-RGE for αv-class integrins.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Fibronectin Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)