Merck
  • Synthesis and antihypertensive effects of new methylthiomorpholinphenol derivatives.
Synthesis and antihypertensive effects of new methylthiomorpholinphenol derivatives.
European journal of medicinal chemistry (2007-06-22)
A Ma Velázquez, L Martínez, V Abrego, M A Balboa, L A Torres, B Camacho, S Díaz-Barriga, A Romero, R López-Castañares, E Angeles
ABSTRACT
We present in this work the synthesis and cardiovascular effects of new methylthiomorpholine compounds and they were compared with cardiovascular drugs such as captopril, losartan and omapatrilat.
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder