Merck
  • Registered report: tumour vascularization via endothelial differentiation of glioblastoma stem-like cells.
Registered report: tumour vascularization via endothelial differentiation of glioblastoma stem-like cells.
eLife (2015-02-26)
Denise Chroscinski, Darryl Sampey, Nimet Maherali
ABSTRACT
The Nature in 2010 (Ricci-Vitiani et al., 2010). The experiments that will be replicated are those reported in Figure 4B and Supplementary Figure 10B (Ricci-Vitiani et al., 2010), which demonstrate that glioblastoma stem-like cells can derive into endothelial cells, and can be selectively ablated to reduce tumor progression in vivo, and Supplementary Figures S10C and S10D (Ricci-Vitiani et al., 2010), which demonstrate that fully differentiated glioblastoma cells cannot form functionally relevant endothelium. The Reproducibility Project: Cancer Biology is a collaboration between the eLife.
Sigma-Aldrich
Ganciclovir, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
First-strand cDNA Synthesis Kit, Cytiva 27-9261-01