Merck
Methods for studying metabolism in Drosophila.
Methods (San Diego, Calif.) (2014-03-19)
Jason M Tennessen, William E Barry, James Cox, Carl S Thummel
ABSTRACT
Recent research using Drosophila melanogaster has seen a resurgence in studies of metabolism and physiology. This review focuses on major methods used to conduct this work. These include protocols for dietary interventions, measurements of triglycerides, cholesterol, glucose, trehalose, and glycogen, stains for lipid detection, and the use of gas chromatography-mass spectrometry (GC-MS) to detect major polar metabolites. It is our hope that this will provide a useful framework for both new and current researchers in the field.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Sudan Black B, certified by the BSC
Sigma-Aldrich
Oil Red O, certified by the BSC
Supelco
Glucose (HK) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides