Merck
Structure-activity relationship of cyanine tau aggregation inhibitors.
Journal of medicinal chemistry (2009-05-13)
Edward Chang, Erin E Congdon, Nicolette S Honson, Karen E Duff, Jeff Kuret
ABSTRACT
A structure-activity relationship for symmetrical cyanine inhibitors of human tau aggregation was elaborated using a filter trap assay. Antagonist activity depended on cyanine heterocycle, polymethine bridge length, and the nature of meso- and N-substituents. One potent member of the series, 3,3'-diethyl-9-methylthiacarbocyanine iodide (compound 11), retained submicromolar potency and had calculated physical properties consistent with blood-brain barrier and cell membrane penetration. Exposure of organotypic slices prepared from JNPL3 transgenic mice (which express human tau harboring the aggregation prone P301L tauopathy mutation) to compound 11 for one week revealed a biphasic dose response relationship. Low nanomolar concentrations decreased insoluble tau aggregates to half those observed in slices treated with vehicle alone. In contrast, high concentrations (> or =300 nM) augmented tau aggregation and produced abnormalities in tissue tubulin levels. These data suggest that certain symmetrical carbocyanine dyes can modulate tau aggregation in the slice biological model at concentrations well below those associated with toxicity.
Sigma-Aldrich
3,3′-Diethylthiadicarbocyanine iodide, Dye content 98 %
Sigma-Aldrich
3,3′-Diethylthiatricarbocyanine iodide, 99%
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
3,3′-Diethylthiacarbocyanine iodide, Dye content 95 %
Sigma-Aldrich
3,3′-Diethyloxacarbocyanine iodide, 98%
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy