Merck
  • Loss of ATRX/DAXX expression and alternative lengthening of telomeres in uterine leiomyomas.
Loss of ATRX/DAXX expression and alternative lengthening of telomeres in uterine leiomyomas.
Cancer (2018-11-14)
Terhi V Ahvenainen, Netta M Mäkinen, Pernilla von Nandelstadh, Maija E A Vahteristo, Annukka M Pasanen, Ralf C Bützow, Pia M Vahteristo
ABSTRACT
Uterine leiomyomas (ULs) are the most common gynecologic tumors and affect 3 of every 4 women by the age of 50 years. The majority of ULs are classified as conventional tumors, whereas 10% represent various histopathological subtypes with features that mimic malignancy. These subtypes include cellular and mitotically active ULs and ULs with bizarre nuclei. Uterine leiomyosarcoma (ULMS), the malignant counterpart of UL, is an aggressive cancer with poor overall survival. The early diagnosis and preoperative differentiation of ULMS from UL are often challenging because their symptoms and morphology resemble one another. Recent studies have shown frequent loss of alpha-thalassemia/mental retardation syndrome X-linked (ATRX) or death domain-associated protein (DAXX) expression in ULMS, and this is often associated with an alternative lengthening of telomeres (ALT) phenotype. To investigate ATRX and DAXX expression and the presence of ALT in UL subtypes, immunohistochemical and telomere-specific fluorescence in situ hybridization analyses were performed. The study material consisted of 142 formalin-fixed, paraffin-embedded tissue samples representing various UL subtypes and 64 conventional ULs. A loss of ATRX or DAXX and/or ALT was detected in 6.3% of the histopathological UL subtype samples (9 of 142). Two patients whose ULs showed either ATRX loss or ALT were later diagnosed with a pulmonary smooth muscle tumor. Pulmonary tumors displayed molecular alterations found in the corresponding uterine tumors, which indicated metastasis to the lungs. All conventional ULs displayed normal ATRX, DAXX, and telomeres. These results highlight the differences between conventional and histopathologically atypical ULs and indicate that some UL subtype tumors may harbor long-term malignant potential.
Sigma-Aldrich
Anti-DAXX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ATRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution