Merck
  • Carbonic anhydrase activators: activation of human isozymes I, II and IX with phenylsulfonylhydrazido l-histidine derivatives.
Carbonic anhydrase activators: activation of human isozymes I, II and IX with phenylsulfonylhydrazido l-histidine derivatives.
Bioorganic & medicinal chemistry letters (2009-04-07)
Marie-Rose Abdo, Daniela Vullo, Mohamed-Chiheb Saada, Jean-Louis Montero, Andrea Scozzafava, Jean-Yves Winum, Claudiu T Supuran
ABSTRACT
Activation of the human carbonic anhydrase (CA, EC 4.2.1.1) isozymes I, II (cytosolic) and IX (transmembrane, tumor-associated isoform) with a series of arylsulfonylhydrazido-l-histidines incorporating 4-substituted-phenyl, pentafluorophenyl- and beta-naphthyl moieties was investigated. The compounds showed a weak hCA I activation profile, but were more efficient as hCA II and IX activators. The 4-iodophenyl-substituted derivative behaved as a strong and isozyme selective hCA II activator, with an activation constant of 0.21muM. This is the first isoform-selective, potent CA activator reported to date.
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland