Merck
  • Tumour-associated macrophages drive stromal cell-dependent collagen crosslinking and stiffening to promote breast cancer aggression.
Tumour-associated macrophages drive stromal cell-dependent collagen crosslinking and stiffening to promote breast cancer aggression.
Nature materials (2020-12-02)
Ori Maller, Allison P Drain, Alexander S Barrett, Signe Borgquist, Brian Ruffell, Igor Zakharevich, Thanh T Pham, Tina Gruosso, Hellen Kuasne, Johnathon N Lakins, Irene Acerbi, J Matthew Barnes, Travis Nemkov, Aastha Chauhan, Jessica Gruenberg, Aqsa Nasir, Olof Bjarnadottir, Zena Werb, Peter Kabos, Yunn-Yi Chen, E Shelley Hwang, Morag Park, Lisa M Coussens, Andrew C Nelson, Kirk C Hansen, Valerie M Weaver
ABSTRACT
Stromal stiffening accompanies malignancy, compromises treatment and promotes tumour aggression. Clarifying the molecular nature and the factors that regulate stromal stiffening in tumours should identify biomarkers to stratify patients for therapy and interventions to improve outcome. We profiled lysyl hydroxylase-mediated and lysyl oxidase-mediated collagen crosslinks and quantified the greatest abundance of total and complex collagen crosslinks in aggressive human breast cancer subtypes with the stiffest stroma. These tissues harbour the highest number of tumour-associated macrophages, whose therapeutic ablation in experimental models reduced metastasis, and decreased collagen crosslinks and stromal stiffening. Epithelial-targeted expression of the crosslinking enzyme, lysyl oxidase, had no impact on collagen crosslinking in PyMT mammary tumours, whereas stromal cell targeting did. Stromal cells in microdissected human tumours expressed the highest level of collagen crosslinking enzymes. Immunohistochemical analysis of biopsies from a cohort of patients with breast cancer revealed that stromal expression of lysyl hydroxylase 2, an enzyme that induces hydroxylysine aldehyde-derived collagen crosslinks and stromal stiffening, correlated significantly with disease specific mortality. The findings link tissue inflammation, stromal cell-mediated collagen crosslinking and stiffening to tumour aggression and identify lysyl hydroxylase 2 as a stromal biomarker.
Product Description
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Anti-phospho SMAD2 Antibody (Ser465/467), from rabbit, purified by affinity chromatography
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks