Merck
  • Glucose-based spiro-isoxazolines: a new family of potent glycogen phosphorylase inhibitors.
Glucose-based spiro-isoxazolines: a new family of potent glycogen phosphorylase inhibitors.
Bioorganic & medicinal chemistry (2009-09-29)
Mahmoud Benltifa, Joseph M Hayes, Sébastien Vidal, David Gueyrard, Peter G Goekjian, Jean-Pierre Praly, Gregory Kizilis, Costas Tiraidis, Kyra-Melinda Alexacou, Evangelia D Chrysina, Spyros E Zographos, Demetres D Leonidas, Georgios Archontis, Nikos G Oikonomakos
ABSTRACT
A series of glucopyranosylidene-spiro-isoxazolines was prepared through regio- and stereoselective [3+2]-cycloaddition between the methylene acetylated exo-glucal and aromatic nitrile oxides. The deprotected cycloadducts were evaluated as inhibitors of muscle glycogen phosphorylase b. The carbohydrate-based family of five inhibitors displays K(i) values ranging from 0.63 to 92.5 microM. The X-ray structures of the enzyme-ligand complexes show that the inhibitors bind preferentially at the catalytic site of the enzyme retaining the less active T-state conformation. Docking calculations with GLIDE in extra-precision (XP) mode yielded excellent agreement with experiment, as judged by comparison of the predicted binding modes of the five ligands with the crystallographic conformations and the good correlation between the docking scores and the experimental free binding energies. Use of docking constraints on the well-defined positions of the glucopyranose moiety in the catalytic site and redocking of GLIDE-XP poses using electrostatic potential fit-determined ligand partial charges in quantum polarized ligand docking (QPLD) produced the best results in this regard.
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Millipore
Dextrin from potato starch, for biotechnological purposes
Millipore
Dextrin from potato starch, suitable for microbiology
Sigma-Aldrich
Dextrin from corn, Type I, powder
Sigma-Aldrich
Dextrin from corn, Type II, powder
Sigma-Aldrich
Glucose solution, BioUltra, Molecular Biology, ~20% in H2O
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications