- Patient-Derived Tumor Organoid Rings for Histologic Characterization and High-Throughput Screening.
Patient-Derived Tumor Organoid Rings for Histologic Characterization and High-Throughput Screening.
STAR protocols (2020-10-13)
Huyen Thi Lam Nguyen, Alice Soragni
PMID33043307
ABSTRACT
Tumor organoids are promising tools for cancer biology investigations and preclinical drug screenings because they are often representative of the histology and drug responses of patients. Here, we introduce a facile protocol to overcome technical limitations by generating patient-derived tumor organoids using a simplified ring-like geometry. This facilitates media exchange and drug treatment for histopathology characterization and automated high-throughput drug screenings. For complete details on the use and execution of this protocol, please refer to Phan et al. (2019).