Merck
  • Structural features of bovine colostral immunoglobulin that confer proteolytic stability in a simulated intestinal fluid.
Structural features of bovine colostral immunoglobulin that confer proteolytic stability in a simulated intestinal fluid.
The Journal of biological chemistry (2020-07-16)
Randall E Burton, Skaison Kim, Rutvij Patel, Deborah S Hartman, Daniel E Tracey, Barbara S Fox
ABSTRACT
Bovine colostral antibodies, purified from cow's milk produced immediately after calving, have enhanced resistance to degradation by intestinal proteases relative to antibodies from human or bovine serum, making them of particular interest as orally administered therapeutic agents. However, the basis of this resistance is not well defined. We evaluated the stability of AVX-470, a bovine colostral anti-tumor necrosis factor (TNF) polyclonal antibody used in early clinical studies for treatment of ulcerative colitis, using conditions that mimic the human small intestine. AVX-470 was degraded ∼3 times more slowly than human IgG antibodies or infliximab (a monoclonal mouse-human chimeric IgG). Bovine IgG1 antibodies, the primary component of AVX-470, were slowly cleaved to F(ab')2 fragments. In contrast, bovine IgG2 and human IgG1 antibodies were cleaved rapidly into Fab and smaller fragments, pointing to specific regions where additional stability might be gained. Infliximab was modified to incorporate the sequences from these regions, including the bovine IgG1 hinge region and a predicted disulfide bonding motif linking the upper hinge region, the CH1 domain, and the light chain. This infliximab-bovine IgG1 chimera (bovinized infliximab) retained the antigen binding and neutralization activity of the WT sequence but was degraded 9-fold more slowly than the unmodified infliximab. This remarkable increase in stability with as few as 18 amino acid substitutions suggests that this bovinization process is a means to enable oral delivery of proven therapeutic antibodies as well as novel antibodies to targets that have been previously inaccessible to therapies delivered by injection.
Sigma-Aldrich
Pancreatin from porcine pancreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%