Merck
  • Patch-Clamp Analysis of the Mitochondrial H+ Leak in Brown and Beige Fat.
Patch-Clamp Analysis of the Mitochondrial H+ Leak in Brown and Beige Fat.
Frontiers in physiology (2020-05-01)
Ambre M Bertholet, Yuriy Kirichok
ABSTRACT
Mitochondria convert the chemical energy of metabolic substrates into adenosine triphosphate (ATP) and heat. Although ATP production has become a focal point of research in bioenergetics, mitochondrial thermogenesis is also crucial for energy metabolism. Mitochondria generate heat due to H+ leak across the inner mitochondrial membrane (IMM) which is mediated by mitochondrial uncoupling proteins. The mitochondrial H+ leak was first identified, and studied for many decades, using mitochondrial respiration technique. Unfortunately, this method measures H+ leak indirectly, and its precision is insufficient for the rigorous insight into the mitochondrial function at the molecular level. Direct patch-clamp recording of H+ leak would have a significantly higher amplitude and time resolution, but application of the patch-clamp technique to a small subcellular organelle such as mitochondria has been challenging. We developed a method that facilitates patch-clamp recording from the whole IMM, enabling the direct measurement of small H+ leak currents via uncoupling proteins and thus, providing a rigorous understanding of the molecular mechanisms involved. In this paper we cover the methodology of measuring the H+ leak in mitochondria of specialized thermogenic tissues brown and beige fat.
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine 5′-diphosphate tris salt from Saccharomyces cerevisiae, Type VI, ≥92.5%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Arachidonic acid sodium salt from Mortierella alpina, ≥98.5% (GC)