Merck
  • Organ-on-chip model shows that ATP release through connexin hemichannels drives spontaneous Ca2+ signaling in non-sensory cells of the greater epithelial ridge in the developing cochlea.
Organ-on-chip model shows that ATP release through connexin hemichannels drives spontaneous Ca2+ signaling in non-sensory cells of the greater epithelial ridge in the developing cochlea.
Lab on a chip (2020-07-24)
Flavia Mazzarda, Annunziata D'Elia, Roberto Massari, Adele De Ninno, Francesca Romana Bertani, Luca Businaro, Gaia Ziraldo, Veronica Zorzi, Chiara Nardin, Chiara Peres, Francesco Chiani, Abraham Tettey-Matey, Marcello Raspa, Ferdinando Scavizzi, Alessandro Soluri, Anna Maria Salvatore, Jun Yang, Fabio Mammano
ABSTRACT
Prior work supports the hypothesis that ATP release through connexin hemichannels drives spontaneous Ca2+ signaling in non-sensory cells of the greater epithelial ridge (GER) in the developing cochlea; however, direct proof is lacking. To address this issue, we plated cochlear organotypic cultures (COCs) and whole cell-based biosensors with nM ATP sensitivity (ATP-WCBs) at the bottom and top of an ad hoc designed transparent microfluidic chamber, respectively. By performing dual multiphoton Ca2+ imaging, we monitored the propagation of intercellular Ca2+ waves in the GER of COCs and ATP-dependent Ca2+ responses in overlying ATP-WCBs. Ca2+ signals in both COCs and ATP-WCBs were inhibited by supplementing the extracellular medium with ATP diphosphohydrolase (apyrase). Spontaneous Ca2+ signals were strongly depressed in the presence of Gjb6-/- COCs, in which connexin 30 (Cx30) is absent and connexin 26 (Cx26) is strongly downregulated. In contrast, spontaneous Ca2+ signals were not affected by replacement of Panx1-/- with Panx1+/+ COCs in the microfluidic chamber. Similar results were obtained by estimating ATP release from COCs using a classical luciferin-luciferase bioluminescence assay. Therefore, connexin hemichannels and not pannexin 1 channels mediate the release of ATP that is responsible for Ca2+ wave propagation in the developing mouse cochlea. The technological advances presented here have the potential to shed light on a plethora of unrelated open issues that involve paracrine signaling in physiology and pathology and cannot be addressed with standard methods.
Product Description
Supelco
(±)-Sulfinpyrazone, analytical standard
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
ARL 67156 trisodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Suramin sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture