Merck
Measurement of ATPase Activity of Valosin-containing Protein/p97.
Bio-protocol (2021-03-04)
Kruthi Suvarna, Kaori Honda, Makoto Muroi, Yasumitsu Kondoh, Hiroyuki Osada, Nobumoto Watanabe
ABSTRACT
Valosin-containing protein (VCP; also known as p97) is a type II ATPase regulating several cellular processes. Using proteomic techniques, we identified a chemical compound that binds to the D1 ATPase domain of VCP. The protocol described here was to study the effect of the compound on ATPase activity in vitro of purified VCP protein. ATPases are enzymes that hydrolyze ATP in a reaction resulting the release of an inorganic phosphate. This reaction can be measured using several methods, such as colorimetric, fluorescence, and radiometric assays, in addition to the bioluminescence assay mentioned here. Since the remaining ATP level after the reaction was detected using a luciferase assay, the luminescent signal indicates the ATPase activity inversely. This protocol is sensitive, rapid, and can be used for high-throughput screening assays to study the effect of compounds on ATPase function.
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
DBeQ, ≥98% (HPLC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
CHAPS hydrate, ≥98% (HPLC)