Merck
  • H-NS modulates multidrug resistance of Salmonella enterica serovar Typhimurium by repressing multidrug efflux genes acrEF.
H-NS modulates multidrug resistance of Salmonella enterica serovar Typhimurium by repressing multidrug efflux genes acrEF.
Antimicrobial agents and chemotherapy (2009-06-10)
Kunihiko Nishino, Mitsuko Hayashi-Nishino, Akihito Yamaguchi
ABSTRACT
Screening of Salmonella mutants for the ability to increase beta-lactam resistance has led to the identification of a mutation in hns, which codes for the histone-like nucleoid structuring protein (H-NS). In this study, we report that H-NS modulates multidrug resistance through repression of the genes that encode the AcrEF multidrug efflux pump in Salmonella enterica serovar Typhimurium.
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O