Merck
  • Staphylococcus aureus-Derived α-Hemolysin Evokes Generation of Specialized Pro-resolving Mediators Promoting Inflammation Resolution.
Staphylococcus aureus-Derived α-Hemolysin Evokes Generation of Specialized Pro-resolving Mediators Promoting Inflammation Resolution.
Cell reports (2020-10-15)
Paul M Jordan, Jana Gerstmeier, Simona Pace, Rossella Bilancia, Zhigang Rao, Friedemann Börner, Laura Miek, Óscar Gutiérrez-Gutiérrez, Vandana Arakandy, Antonietta Rossi, Armando Ialenti, Cristina González-Estévez, Bettina Löffler, Lorena Tuchscherr, Charles N Serhan, Oliver Werz
ABSTRACT
Underlying mechanisms of how infectious inflammation is resolved by the host are incompletely understood. One hallmark of inflammation resolution is the activation of specialized pro-resolving mediators (SPMs) that enhance bacterial clearance and promote tissue repair. Here, we reveal α-hemolysin (Hla) from Staphylococcus aureus as a potent elicitor of SPM biosynthesis in human M2-like macrophages and in the mouse peritoneum through selective activation of host 15-lipoxygenase-1 (15-LOX-1). S. aureus-induced SPM formation in M2 is abolished upon Hla depletion or 15-LOX-1 knockdown. Isolated Hla elicits SPM formation in M2 that is reverted by inhibition of the Hla receptor ADAM10. Lipid mediators derived from Hla-treated M2 accelerate planarian tissue regeneration. Hla but not zymosan provokes substantial SPM formation in the mouse peritoneum, devoid of leukocyte infiltration and pro-inflammatory cytokine secretion. Besides harming the host, Hla may also exert beneficial functions by stimulating SPM production to promote the resolution of infectious inflammation.
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.
Sigma-Aldrich
GI254023X, ≥98% (HPLC)
Sigma-Aldrich
α-Hemolysin from Staphylococcus aureus, lyophilized powder, Protein ~60 % by Lowry, ≥10,000 units/mg protein
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077