Merck
Integrative Modeling of a Sin3/HDAC Complex Sub-structure.
Cell reports (2020-04-16)
Charles A S Banks, Ying Zhang, Sayem Miah, Yan Hao, Mark K Adams, Zhihui Wen, Janet L Thornton, Laurence Florens, Michael P Washburn
ABSTRACT
Sin3/HDAC complexes function by deacetylating histones, condensing chromatin, and modulating gene expression. Although components used to build these complexes have been well defined, we still have only a limited understanding of the structure of the Sin3/HDAC subunits assembled around the scaffolding protein SIN3A. To characterize the spatial arrangement of Sin3 subunits, we combined Halo affinity capture, chemical crosslinking, and high-resolution mass spectrometry (XL-MS) to determine intersubunit distance constraints, identifying 66 interprotein and 63 self-crosslinks for 13 Sin3 subunits. Having assessed crosslink authenticity by mapping self-crosslinks onto existing structures, we used distance restraints from interprotein crosslinks to guide assembly of a Sin3 complex substructure. We identified the relative positions of subunits SAP30L, HDAC1, SUDS3, HDAC2, and ING1 around the SIN3A scaffold. The architecture of this subassembly suggests that multiple factors have space to assemble to collectively influence the behavior of the catalytic subunit HDAC1.
Sigma-Aldrich
2-Chloroacetamide, ≥98%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
1,10-Phenanthroline monohydrate, reagent grade
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)