Merck
  • DNA Fiber Assay for the Analysis of DNA Replication Progression in Human Pluripotent Stem Cells.
DNA Fiber Assay for the Analysis of DNA Replication Progression in Human Pluripotent Stem Cells.
Current protocols in stem cell biology (2020-06-26)
Jason A Halliwell, Polly Gravells, Helen E Bryant
ABSTRACT
Human pluripotent stem cells (PSC) acquire recurrent chromosomal instabilities during prolonged in vitro culture that threaten to preclude their use in cell-based regenerative medicine. The rapid proliferation of pluripotent cells leads to constitutive replication stress, hindering the progression of DNA replication forks and in some cases leading to replication-fork collapse. Failure to overcome replication stress can result in incomplete genome duplication, which, if left to persist into the subsequent mitosis, can result in structural and numerical chromosomal instability. We have recently applied the DNA fiber assay to the study of replication stress in human PSC and found that, in comparison to somatic cells states, these cells display features of DNA replication stress that include slower replication fork speeds, evidence of stalled forks, and replication initiation from dormant replication origins. These findings have expanded on previous work demonstrating that extensive DNA damage in human PSC is replication associated. In this capacity, the DNA fiber assay has enabled the development of an advanced nucleoside-enriched culture medium that increases replication fork progression and decreases DNA damage and mitotic errors in human PSC cultures. The DNA fiber assay allows for the study of replication fork dynamics at single-molecule resolution. The assay relies on cells incorporating nucleotide analogs into nascent DNA during replication, which are then measured to monitor several replication parameters. Here we provide an optimized protocol for the fiber assay intended for use with human PSC, and describe the methods employed to analyze replication fork parameters. © 2020 Wiley Periodicals LLC. Basic Protocol 1: DNA fiber labeling Basic Protocol 2: DNA fiber spreading Basic Protocol 3: Immunostaining Support Protocol 1: Microscopy/data acquisition Support Protocol 2: Data analysis.
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
5-Iodo-2′-deoxyuridine, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Fluoroshield, histology mounting medium
Pricing and availability is not currently available.
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog
Pricing and availability is not currently available.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.