Merck
  • Chemical genomics in Escherichia coli identifies an inhibitor of bacterial lipoprotein targeting.
Chemical genomics in Escherichia coli identifies an inhibitor of bacterial lipoprotein targeting.
Nature chemical biology (2009-09-29)
Ranjana Pathania, Soumaya Zlitni, Courtney Barker, Rahul Das, David A Gerritsma, Julie Lebert, Emilia Awuah, Giuseppe Melacini, Fred A Capretta, Eric D Brown
ABSTRACT
One of the most significant hurdles to developing new chemical probes of biological systems and new drugs to treat disease is that of understanding the mechanism of action of small molecules discovered with cell-based small-molecule screening. Here we have assembled an ordered, high-expression clone set of all of the essential genes from Escherichia coli and used it to systematically screen for suppressors of growth inhibitory compounds. Using this chemical genomic approach, we demonstrate that the targets of well-known antibiotics can be identified as high copy suppressors of chemical lethality. This approach led to the discovery of MAC13243, a molecule that belongs to a new chemical class and that has a unique mechanism and promising activity against multidrug-resistant Pseudomonas aeruginosa. We show that MAC13243 inhibits the function of the LolA protein and represents a new chemical probe of lipoprotein targeting in bacteria with promise as an antibacterial lead with Gram-negative selectivity.
Product Description
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
D-Cycloserine, synthetic
Sigma-Aldrich
D-Cycloserine
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Sulfamethoxazole
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Norfloxacin, VETRANAL®, analytical standard