Merck
  • Acrylamide and its metabolite induce neurotoxicity via modulation of protein kinase C and AMP-activated protein kinase pathways.
Acrylamide and its metabolite induce neurotoxicity via modulation of protein kinase C and AMP-activated protein kinase pathways.
Toxicology in vitro : an international journal published in association with BIBRA (2021-02-06)
Dahlia Triningsih, Jae-Ho Yang, Kyeong Hwa Sim, Chuhee Lee, Youn Ju Lee
ABSTRACT
Acrylamide is known as a neurotoxicant found in commonly consumed food as well as in human body. However, the underlying mechanisms involved in neurotoxicity by acrylamide and its metabolite, glycidamide remain largely unknown. In this study, we have examined the interplay between CYP2E1, AMPK, ERK and PKC in acrylamide-induced neurotoxicity associated with autophagy in PC12 cells. Acrylamide-induced cell death was mediated by CYP2E1 expression and the activation of ERK, PKC-ɑ and PKC-δ, whereas AMPK knockdown exacerbated the acrylamide-induced neurotoxic effects. PKC-ɑ, but not PKC-δ, plays an upstream regulator of ERK and AMPK. Moreover, AMPK activation suppressed ERK, and CYP2E1 and AMPK bilaterally inhibit each other. Furthermore, acrylamide increased autophagy with impaired autophagic flux, evidenced by the increased beclin-1, LC3-II and p62 protein. Acrylamide-induced neuronal death was ameliorated by 3-methyladenine, an autophagy inhibitor, whereas neuronal death was exacerbated by chloroquine, a lysosomal inhibitor. Interestingly, PKC-δ siRNA, but not PKC-ɑ siRNA, dramatically reduced acrylamide-induced beclin-1 and LC3-II levels, whereas AMPK siRNA further increased beclin-1, LC3-II and p62 protein levels. Glycidamide, a major metabolite, mimicked acrylamide only with a higher potency. Taken together, acrylamide- and glycidamide-induced neurotoxicity may involve cytotoxic autophagy, which is mediated by interplay between PKCs and AMPK pathways.
Product Description
Supelco
Diacetoxyscirpenol solution, ~100 μg/mL in acetonitrile, analytical standard
Supelco
Glycidamide, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human CYP2E1
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder