Merck
  • N-((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)arylamide as a new scaffold that provides rapid access to antimicrotubule agents: synthesis and evaluation of antiproliferative activity against select cancer cell lines.
N-((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)arylamide as a new scaffold that provides rapid access to antimicrotubule agents: synthesis and evaluation of antiproliferative activity against select cancer cell lines.
Journal of medicinal chemistry (2010-03-26)
Jonathan A Stefely, Rahul Palchaudhuri, Patricia A Miller, Rebecca J Peterson, Garrett C Moraski, Paul J Hergenrother, Marvin J Miller
ABSTRACT
A series of N-((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)arylamides was synthesized by copper-catalyzed azide-alkyne cycloaddition (CuAAC) and afforded inhibitors of cancer cell growth. For example, compound 13e had an IC(50) of 46 nM against MCF-7 human breast tumor cells. Structure-activity relationship (SAR) studies demonstrated that (i) meta-phenoxy substitution of the N-1-benzyl group is important for antiproliferative activity and (ii) a variety of heterocyclic substitutions for the aryl group of the arylamide are tolerated. In silico COMPARE analysis of antiproliferative activity against the NCI-60 human tumor cell line panel revealed a correlation to clinically useful antimicrotubule agents such as paclitaxel and vincristine. This in silico correlation was supported by (i) in vitro inhibition of tubulin polymerization, (ii) G(2)/M-phase arrest in HeLa cells as assessed by flow cytometry, and (iii) perturbation of normal microtubule activity in HeLa cells as observed by confocal microscopy. The results demonstrate that N-((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)arylamide is a readily accessible small molecule scaffold for compounds that inhibit tubulin polymerization and tumor cell growth.
Sigma-Aldrich
2-Methoxyestradiol, powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder